Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
pH 4.65 DIN 19267 Buffer Solution at 25oC 1 * 500 mL
Catalog # REGE104655
Supplier:
REAGECON
New Product
Expand image
pH 4.65 DIN 19267 Buffer Solution at 25oC 1 * 500 mL
Catalog # REGE104655
Supplier:
REAGECON
New Product