Pepsin
Poly bottle, 500 g
Catalog # SPCMP1571-500GM
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Commonly used in biochemistry.
Specifications
Specifications
- Pack type:Poly bottle
- Pk:500 g
- Cat. No.:SPCMP1571-500GM
- Form:Powder
- Storage conditions:Refrigerator
- Grade:EP
- Tested applications:for biochemistry
- Enzyme name:Pepsin
- CAS No.:9001-75-6
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



