Order Entry
Finland
Orders LinkContactUsLinkComponent
Pepsin

Pepsin

Poly bottle, 100 g
Catalog # SPCMP1571-100GM
Small Business Enterprise
Supplier:   Spectrum Chemical
Pepsin
Catalog # SPCMP1571-100GM
Supplier:  Spectrum Chemical
Small Business Enterprise
CAS Number:  
Pepsin

Poly bottle, 100 g

CAS Number:  
Price
405,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Commonly used in biochemistry.

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    100 g
  • Cat. No.:
    SPCMP1571-100GM
  • Form:
    Powder
  • Storage conditions:
    Refrigerator
  • Grade:
    EP
  • Tested applications:
    for biochemistry
  • Enzyme name:
    Pepsin
  • CAS No.:
    9001-75-6

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.