Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Mouse Recombinant TNF-alpha

1.000 µG
Catalog # STMC78069.3
Marketsource
Mouse Recombinant TNF-alpha
Catalog # STMC78069.3
Marketsource
CAS Number:  
Mouse Recombinant TNF-alpha
Mouse Recombinant TNF-alpha

1.000 µG

CAS Number:  
Price
4 330,00 €
Log in for availability
1 * 1 Each
Product Details & Documents

Tumor necrosis factor-α (TNF-α) is a pro-inflammatory cytokine that activates NF-κB, MAPK, and PI3K/AKT pathways. Activated T cells and macrophages are the primary producers of TNF-α in response to inflammation and infectious conditions. Many other cell types have been shown to produce TNF-α, among them B cells, NK cells, mast cells, neutrophils, dendritic cells, microglia, endothelial cells, smooth muscle cells, cardiomyocytes, and fibroblasts. TNF-α has cytotoxic effects on cancer cells in vitro by stimulating anti-tumor immunosuppressive responses. TNF-α stimulates expression of E- and P-selectins, thus facilitating adhesion of neutrophils, monocytes, and memory T cells to activated platelets and endothelial cells (Zelová andamp; Hosek). Other effects of TNF-α include vasodilatation and edema formation.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    1.000 µG
  • Cat. No.:
    STMC78069.3
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    TNF-alpha
  • Purity:
    ≥95%
  • Species:
    Mouse

Specifications