Order Entry
Finland
Orders LinkContactUsLinkComponent
4'-Fluoroacetophenone 95%

4'-Fluoroacetophenone 95%

Bottle, 500 g
Catalog # APOSPC2887-500G
Irritant
Supplier:   Apollo Scientific
4'-Fluoroacetophenone 95%
Catalog # APOSPC2887-500G
Supplier:  Apollo Scientific
CAS Number:  403-42-9
4'-Fluoroacetophenone 95%

Bottle, 500 g

CAS Number:  403-42-9
Price
51,50 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
  • CAS Number:403-42-9
  • Formula:C₈H₇FO
  • MDL Number:MFCD00000354
  • Molecular Weight:138.14 g/mol
  • Boiling Pt:194…196 °C
  • Melting Pt:4 °C
  • Density:1.138 g/cm³ (25 °C)
  • Flash Pt:71 °C
  • Storage Temperature:Ambient
  • EINECS:206-960-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications