Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Test Sieve ISO 3310/2 200x50mm 4.75mm stainless steel perforated plate, square hole 1 * 1 items
Catalog # RETS60.190.004750
Supplier:
Retsch GmbH
Expand image
Test Sieve ISO 3310/2 200x50mm 4.75mm stainless steel perforated plate, square hole 1 * 1 items
Catalog # RETS60.190.004750
Supplier:
Retsch GmbH