ACE-2
Plastic bottle, 10 mg
Catalog # AATB13556
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
ACE2 (angiotensin-converting enzyme 2) is a metalloproteinase that requires a divalent cation positioned at the active site in order to perform catalysis.
- High flexibility
- Higher throughput
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:10 mg
- Cat. No.:AATB13556
- Molecular formula:C₃₂H₃₆N₆O₁₂
- Protein/peptide name:ACE-2
- Grade:Molecular Biology Grade
- Tested applications:Fluorescence
- Molecular weight:696,7 g/mol
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



