Order Entry
Finland
ContactUsLinkComponent
Support frame, for under bench cabinet, width 590 mm, sheet steel powder-coated / panel melamine resin-coated, RAL 7035 light grey for hazardous materials work stations with width 1200 mm with depth 750 mm , for model(s): GAP Version: SV version - st 1 * 1 items
Catalog # MSGA41074
Supplier:  asecos
Support frame, for under bench cabinet, width 590 mm, sheet steel powder-coated / panel melamine resin-coated, RAL 7035 light grey for hazardous materials work stations with width 1200 mm with depth 750 mm , for model(s): GAP Version: SV version - st 1 * 1 items
Catalog # MSGA41074
Supplier:  asecos
Frequently Bought Together