Order Entry
Finland
ContactUsLinkComponent
Support frame, for 1400 mm wide under bench cabinet, steel powder-coated / panel melamine resin-coated, RAL 7035 light grey for hazardous materials work stations with width 1800 mm with depth 600 mm, for model(s): GAP Version: SV version - standing w 1 * 1 items
Catalog # MSGA41095
Supplier:  asecos
Support frame, for 1400 mm wide under bench cabinet, steel powder-coated / panel melamine resin-coated, RAL 7035 light grey for hazardous materials work stations with width 1800 mm with depth 600 mm, for model(s): GAP Version: SV version - standing w 1 * 1 items
Catalog # MSGA41095
Supplier:  asecos
Frequently Bought Together