Chrysoeriol
Diethylene glycol cyclic phthalate, 236.22, 1 g, 1 g
Catalog # PROOTRCD444310-1G
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 g
- Description:Diethylene glycol cyclic phthalate
- Molecular weight (Mw):236.22
- Volume:1 g
- Cat. No.:PROOTRCD444310-1G
- Formula:C₁₂H₁₂O₅
- Storage temperature:4 °C
- CAS No.:13988-26-6
- Shipping conditions:RT



