Chrysoeriol
Diethylene glycol cyclic phthalate, 236.22, 10 g, 10 g
PROOTRCD444310-10G
- Pk:10 g
- Description:Diethylene glycol cyclic phthalate
- Molecular weight (Mw):236.22
- Volume:10 g
- Cat. No.:PROOTRCD444310-10G
- Formula:C₁₂H₁₂O₅
- Storage temperature:4 °C
- CAS No.:13988-26-6
- Shipping conditions:RT



