Forceps, pointed
Straight, 115 mm
Catalog # COMC200-0115
Supplier: COMACO HANDEL
Jump to:
Product Details & Documents
Stainless steel, grade 18-8.
- Pointed
- With serrated grip and tips
Specifications
- Type:Straight
- Length:115 mm
- Seq:02