Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Grinding Bowls EasyFit PM 300 Agate, 125ml 1 * 1 items
Catalog # RETS01.462.0515
Supplier:
Retsch GmbH
Expand image
Grinding Bowls EasyFit PM 300 Agate, 125ml 1 * 1 items
Catalog # RETS01.462.0515
Supplier:
Retsch GmbH