Order Entry
Finland
ContactUsLinkComponent
SpectraMax iD5 TRF Enhancement Module for enhanced TRF reads including TRF, HTRFÂ, TR-FRET, and enhanced ScanLater western blot dete ction. User-installable. 1 * 1 items
Catalog # MLDV0200-7030-TR
Supplier:  MOLECULAR DEVICES
SpectraMax iD5 TRF Enhancement Module for enhanced TRF reads including TRF, HTRFÂ, TR-FRET, and enhanced ScanLater western blot dete ction. User-installable. 1 * 1 items
Catalog # MLDV0200-7030-TR
Supplier:  MOLECULAR DEVICES