Order Entry
Finland
ContactUsLinkComponent
Canister 10 liter, S60/61, PE-HD electrostatic conductive, with sensor housing for M30 sensorNote: Canisters are marked with prod uc tion date (month / year) and will normally be delivered by us within 6 months from the production date. 1 * 1 items
Catalog # SCTV108292
Supplier:  S.C.A.T.
undefined
Canister 10 liter, S60/61, PE-HD electrostatic conductive, with sensor housing for M30 sensorNote: Canisters are marked with prod uc tion date (month / year) and will normally be delivered by us within 6 months from the production date. 1 * 1 items
Catalog # SCTV108292
Supplier:  S.C.A.T.