Order Entry
Finland
ContactUsLinkComponent
CERT KIT VIAL 0.3ML CLEAR SCREW 9MM SHORT THREAD ID PATCH INSERT CAP SCREW 9MM GREY PP CLEAR SILICONE / CLEAR PTFE SEPTA BONDED SLIT 1.0MM 100/PACK 1 * 100 items
Catalog # HYPE6PCK71LVW
CERT KIT VIAL 0.3ML CLEAR SCREW 9MM SHORT THREAD ID PATCH INSERT CAP SCREW 9MM GREY PP CLEAR SILICONE / CLEAR PTFE SEPTA BONDED SLIT 1.0MM 100/PACK 1 * 100 items
Catalog # HYPE6PCK71LVW
Frequently Bought Together