40x and 60x water immersion system includes 40x apo lwd 1.15 na and 60x plan apo 1.20 na water immersion objectives and dispenser mo dule 1 * 1 items
Catalog # MLDV5076627
40x and 60x water immersion system includes 40x apo lwd 1.15 na and 60x plan apo 1.20 na water immersion objectives and dispenser mo dule 1 * 1 items
Catalog # MLDV5076627