Order Entry
Finland
ContactUsLinkComponent
Oxoid Legionella GVPC Selective Supplement is used for the isolation of Legionella spp. from environmental samples. 1 * 10 Vial
Catalog # OXOISR0252E
Oxoid Legionella GVPC Selective Supplement is used for the isolation of Legionella spp. from environmental samples. 1 * 10 Vial
Catalog # OXOISR0252E