Order Entry
Finland
ContactUsLinkComponent
Fitting, LDPE, Straight, Hosebarb x Quick-Disconnect Couplers, 3/16" ID x 5/16" ID; 100/Pk
Fitting, LDPE, Straight, Hosebarb x Quick-Disconnect Couplers, 3/16" ID x 5/16" ID; 100/Pk
Catalog # MFLX06290-09BLK
undefined
Fitting, LDPE, Straight, Hosebarb x Quick-Disconnect Couplers, 3/16" ID x 5/16" ID; 100/Pk
Catalog # MFLX06290-09BLK
About this item
Provides easy single-handed disconnection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/16" ID x 5/16" ID
Construction from low-density polyethylene (LDPE).
  • Barb size/tubing int.Ø (port 1):
    3/16"
  • Connection Type:
    Quick-Disconnect to Barb
  • Flow pattern:
    Straight
  • Description:
    Fitting, LDPE, Straight, Hosebarb x Quick-Disconnect Couplers, 3/16" ID x 5/16" ID; 100/Pk
  • Seq:
    02
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    100
  • Fitting material:
    LDPE