Acetic acid (glacial) 100% for analysis EMPARTA ACS 1 * 25 L
Catalog # 1.01830.9025
Supplier: Merck
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Pyrogallol for synthesis, Sigma-Aldrich®
Supplier: Merck
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
Ethylene glycol 99.5% for analysis
Supplier: Thermo Fisher Scientific
1-Butanol 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
n-Hexane, anhydrous ≥96.0% (by GC)
Supplier: TCI
1-Butanol for HPLC
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol 70 % pure 1 * 1 L
Supplier: PanReac AppliChem