Order Entry
Finland
Orders LinkContactUsLinkComponent
RBB Buffers

RBB Buffers

RBB Buffer, 4000 ml, 4.000 mL
Catalog # OMEGRBB-4000
Supplier:   OMEGA BIO-TEK
RBB Buffers
Catalog # OMEGRBB-4000
Supplier:  OMEGA BIO-TEK
CAS Number:  
RBB Buffers

RBB Buffer, 4000 ml, 4.000 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 4.000 mL
Product Details & Documents

Caution: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Pk:
    4.000 mL
  • Description:
    RBB Buffer, 4000 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.