Sodium acetate trihydrate EMPROVE API Ph Eur,BP,JP,USP,FCC,E 262 1 * 24 items
Catalog # 1.28204.8336
Supplier: MERCK PRODUCTION CHEMICALS
Sodium acetate trihydrate EMPROVE API Ph Eur,BP,JP,USP,FCC,E 262 1 * 24 items
Catalog # 1.28204.8336
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Potassium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium iodide ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Zinc chloride 98.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Potassium chloride, Suprapur® for trace analysis, Supelco®
Supplier: Merck
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ammonium heptamolybdate tetrahydrate, Supelco®
Supplier: Merck
Calcium chloride, anhydrous ≥96% ACS
Supplier: VWR Chemicals
Sodium carbonate 99.95-100.05% ACS primary standard
Supplier: Thermo Fisher Scientific
Silver nitrate ≥99%
Supplier: Thermo Fisher Scientific
EDTA magnesium dipotassium salt 97%
Supplier: Thermo Fisher Scientific
Sodium periodate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals