Order Entry
Finland
ContactUsLinkComponent
HPLC/GC certified Kit: 548-0030K: 1.5ml Short Thread Vial, ND9, 32 x 11.6mm, amber glass, 1st hydrolytic class, wide opening, label and filling lines + 548-0085K: Screw Cap, ND9, open Top, Polypropylene, blue, liner material: white Silicone/ red PTF 1 * 1.000 items
Catalog # 548-1489A
Supplier:  VWR Collection
HPLC/GC certified Kit: 548-0030K: 1.5ml Short Thread Vial, ND9, 32 x 11.6mm, amber glass, 1st hydrolytic class, wide opening, label and filling lines + 548-0085K: Screw Cap, ND9, open Top, Polypropylene, blue, liner material: white Silicone/ red PTF 1 * 1.000 items
Catalog # 548-1489A
Supplier:  VWR Collection