Order Entry
Finland
ContactUsLinkComponent
Reagecon Hydrochloric Acid Concentrated Volumetric Solution Ampoule to make 1L of 0.1M (0.1N) Hydrochloric Acid UOM 1x Ampoule 1 * 1 Ampoul
Catalog # REGEHC0101L
Supplier:  REAGECON
Reagecon Hydrochloric Acid Concentrated Volumetric Solution Ampoule to make 1L of 0.1M (0.1N) Hydrochloric Acid UOM 1x Ampoule 1 * 1 Ampoul
Catalog # REGEHC0101L
Supplier:  REAGECON