Micropump Gear Pump Service Kit for 74445-15; 1/Ea
Catalog # MFLX74445-93
Supplier: Avantor Fluid Handling
About this item
Extend the life of your gear pump head with these service kits.
- Optimize performance of your Masterflex pumps
Easy to install, these service kits avoid costly downtime of your gear pump head.
Service kit includes: gears, bushings, and seals.
- Description:Gear Pump Service Kit, for 74445-15
- Environmentally Preferable:
- Warranty:1 year
- Pk:1