Order Entry
Finland
ContactUsLinkComponent
Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 3/4" NPT(F) X 3/4" ID
Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 3/4" NPT(F) X 3/4" ID
Catalog # MFLX34031-00
CE certificate
undefined
Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 3/4" NPT(F) X 3/4" ID
Catalog # MFLX34031-00
CE certificate
About this item
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
  • Barb size/tubing int.Ø (port 2):
    3/4"
  • Connection Type:
    Thread to Barb
  • Flow pattern:
    Straight
  • Thread size NPT(F) (port 1):
    3/4"
  • Description:
    Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 3/4" NPT(F)×3/4" ID
  • Environmentally Preferable:
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Warranty:
    2 years
  • Pk:
    1
  • Fitting material:
    PTFE