Masterflex® Replacement Drum Pump Tubes, Avantor®
Drum Pump Tube Set, PP, 47"L
Tuotenumero MFLX70604-44
Toimittaja: Avantor Fluid Handling
Masterflex® Replacement Drum Pump Tubes, Avantor®
Tuotenumero MFLX70604-44
Toimittaja: Avantor Fluid Handling
Jump to:
Product Details & Documents
Safely transfer corrosive chemicals, flammable liquids, and non-corrosive media.
- Heavy-duty, thick-walled construction makes tube more durable
Replacement tubes do not include barrel adapter and mouting bracket; order separately.
Specifications
- Description:Drum Pump Tube Set, PP, 47"L
- Seq:02
- Cat. No.:MFLX70604-44
- Warranty:4 months
- Tube length, inlet:47"
- Model:70604-44
- Wetted materials:Polypropylene