Order Entry
Finland
ContactUsLinkComponent
Micropump 80977 Gear Pump Accessory Service Kit for 07002-20; 1/Ea
Micropump 80977 Gear Pump Accessory Service Kit for 07002-20; 1/Ea
Catalog # MFLX07002-10
undefined
Micropump 80977 Gear Pump Accessory Service Kit for 07002-20; 1/Ea
Catalog # MFLX07002-10
About this item
Extend the life of your gear pump head with these service kits.
  • Optimize performance of your Masterflex pumps
Easy to install, these service kits avoid costly downtime of your gear pump head.
  • Description:
    Gear Pump Service Kit, for 07002-20
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1