Order Entry
Finland
ContactUsLinkComponent
Burkle 5606-0601 Hand Operated Drum Pump with Discharge Tubing, PTFE/FEP, 60 cm inlet tube, 270 mL/stroke
Burkle 5606-0601 Hand Operated Drum Pump with Discharge Tubing, PTFE/FEP, 60 cm inlet tube, 270 mL/stroke
Catalog # MFLX06043-97
undefined
Burkle 5606-0601 Hand Operated Drum Pump with Discharge Tubing, PTFE/FEP, 60 cm inlet tube, 270 mL/stroke
Catalog # MFLX06043-97

About this item

Safely transfer aggressive chemicals.

  • PTFE and FEP wetted materials withstand harsh chemicals
  • Stopcock contains hazardous vapors and prevents dripping

These PTFE barrel pumps are resistant to strong acids, bases, organics, and other chemicals (except elemental fluorine) and are equally suited to ultrapure liquid handling. Choose between pumps with a spout and a discharge hose, depending on your application. All models include a stopcock and require an adapter for use (order separately).

  • Flow rate:
    3,7 strokes/L (14 strokes/gal.)
  • Description:
    Hand Operated Drum Pump with Discharge Tubing, PTFE/FEP, 60 cm inlet tube, 270 mL/stroke
  • Environmentally Preferable:
  • Wetted materials:
    PTFE/FEP
  • Outlet connection:
    Discharge tubing
  • Tube ext.Ø, inlet:
    3,2 cm (1,25")
  • Tube length, inlet:
    60 cm (23,622")
  • Tube length, outlet:
    120 cm (47,25")
  • Warranty:
    15 Months
  • Pk:
    1