Manual Pinch Valve, 7/16" to 1/2" tube OD
Catalog # MFLX98002-20
Supplier: Avantor Fluid Handling
About this item
An ideal option to secure tubing.
- Constructed of durable Delrin®
Fluid contacts only the tubing—ideal for use in pharmaceutical and food industries where eliminating contamination is critical. Obtain precise and repeatable flow rates using easy-to-read metered scale on valve body. Valves are made of Delrin®. Tubing is not included—order flexible tubing with durometer (Shore A) of 60 or less from our "Tubing" section.
- Connection Type:Pinch Valve for Tubing
- Description:Manual Pinch Valve, 7/16" to 1/2" tube OD
- Environmentally Preferable:
- Pinch valve tube size:7/16" to 1/2" OD
- Warranty:3 Months
- Pk:1