Order Entry
Finland
Orders LinkContactUsLinkComponent
Masterflex® Drum and Container Pump Accessories, Avantor®
Masterflex® Drum and Container Pump Accessories, Avantor®
Strainer for Drum Pumps, PP, PP
Catalog # MFLX70604-86
Masterflex® Drum and Container Pump Accessories, Avantor®
Catalog # MFLX70604-86
Masterflex® Drum and Container Pump Accessories, Avantor®
Masterflex® Drum and Container Pump Accessories, Avantor®
Strainer for Drum Pumps, PP, PP
Price
213,00 €
Log in for availability
1 * 1 Items
Product Details & Documents
Extend the life of your drum pump!
  • Optimize performance of your Masterflex® Drum and Container Pumps
Replacement parts are compatible with Masterflex® Complete Drum and Container Pumps, 70613-series.
Specifications
  • Description:
    Strainer for Drum Pumps, PP
  • Material:
    PP
  • Cat. No.:
    MFLX70604-86
  • Warranty:
    4 months