High-Precision High-Pressure Piston Pump Seal Kit, Organic Seal Kit
MFLX41010-76
Designed for optimal operation in your Ismatec pumps.
- Choose from a wide range of replacement parts and accessories
The overall length of Solva 95605-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
- Description:Piston Pump Seal Kit, Organic Seal Kit
- Material:Modified PTFE and fluoroelastomer
- Environmentally Preferable:
- Compatible with:Organic PTFE/Fluoroelastomer Piston Pump with 10 mL/min, 0,13 inlet diameter
- Pk:1