Order Entry
Finland
ContactUsLinkComponent
Masterflex® L/S® Easy-Load® II 2-Channel Pump Head Tubing Retainer Kit for Precision Tubing
Masterflex® L/S® Easy-Load® II 2-Channel Pump Head Tubing Retainer Kit for Precision Tubing
Catalog # MFLX77200-94
undefined
Masterflex® L/S® Easy-Load® II 2-Channel Pump Head Tubing Retainer Kit for Precision Tubing
Catalog # MFLX77200-94
About this item
Enhance the life of your pump head with the replacement retainer kits.
  • Retainer kits provide the parts necessary to replace the spring-loaded tubing retainers in your Masterflex® Easy-Load® II pump heads
  • Suitable for use with Easy-Load® II pump heads 77202-50 and -60
Replacement parts to keep your pump head operating even longer. Tubing retainers hold the tubing in place within the pump head. Pump head sold separately.
Delivery: Includes two small upper retainers, two small lower retainers, and two compression springs.
  • Description:
    Replacement Tubing Retainer Kit for L/S Easy-Load II Pump Heads 77202-50 and -60
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1