Order Entry
Finland
ContactUsLinkComponent
FMI Adapter for 1/4-28 Threaded ID Tubing, Fluoropolymer
FMI Adapter for 1/4-28 Threaded ID Tubing, Fluoropolymer
Catalog # MFLX07115-44
1 year warranty
undefined
FMI Adapter for 1/4-28 Threaded ID Tubing, Fluoropolymer
Catalog # MFLX07115-44
1 year warranty
About this item
Ideal adapters for plastic case pump heads.
  • Optimize performance of your FMI RH Series Piston Pumps
Adapters for your FMI RH series piston pump heads.
  • Description:
    Adapter for 1/4-28 Threaded ID Tubing, Fluoropolymer
  • Environmentally Preferable:
  • Wetted materials:
    Fluoropolymer
  • Warranty:
    1 year
  • Pk:
    1