Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Centrifuge tube according to Kolmer, boro 3.3 glass, 10 ml 0-4 in 1/10, 4-10 in 1/10, with rim, blue graduation 1 * 1 items
Catalog # NEUB0461-80-17125
Supplier:
NEUBERT VOLUME GLASSWAERE
Expand image
Centrifuge tube according to Kolmer, boro 3.3 glass, 10 ml 0-4 in 1/10, 4-10 in 1/10, with rim, blue graduation 1 * 1 items
Catalog # NEUB0461-80-17125
Supplier:
NEUBERT VOLUME GLASSWAERE