Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PLATE, UV CONVERTER, UVP VISI-BLUE 16.8cm X 21cm 1 * 1 items
Catalog # UVPA38-0409-01
Supplier:
UVP ULTRA VIOLET PRODUCTS
Expand image
PLATE, UV CONVERTER, UVP VISI-BLUE 16.8cm X 21cm 1 * 1 items
Catalog # UVPA38-0409-01
Supplier:
UVP ULTRA VIOLET PRODUCTS