Order Entry
Finland
Orders LinkContactUsLinkComponent
VWR®, Trypsin-EDTA solutions

VWR®, Trypsin-EDTA solutions

Trypsin-EDTA 10X, Liquid, 100 ml
Catalog # 392-0459
Supplier:   VWR Chemicals
VWR®, Trypsin-EDTA solutions
Catalog # 392-0459
Supplier:  VWR Chemicals
CAS Number:  
VWR®, Trypsin-EDTA solutions
VWR®, Trypsin-EDTA solutions

Trypsin-EDTA 10X, Liquid, 100 ml

CAS Number:  
Price
16,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

VWR® Trypsin-EDTA solutions are used in cell culture to remove adherent cells from a culture surface. Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to separate cells from their substrate is dependent primarily on the cell type and the age of the culture. EDTA, a chelating agent, enhances enzymatic activity by removing calcium and magnesium ions. These ions obscure the peptide bonds on which trypsin acts as well as enhancing cell to cell adhesion.

  • Trypsin EDTA solutions exist in different buffers and concentrations
  • Trypsin 0,25% - EDTA in HBSS without calcium, without magnesium, with phenol red
  • Trypsin EDTA 1X in PBS without calcium, without magnesium without phenol red
  • Trypsin EDTA 1X without calcium, without magnesium with phenol red
  • Trypsin-EDTA 10X

After treatment, trypsin activity can be stopped by adding a serum media mixture or a trypsin inhibitor.

Specifications

Specifications

  • Pk:
    100 ml
  • Description:
    Trypsin-EDTA 10X
  • Form:
    Liquid
  • Cat. No.:
    392-0459
  • Sterility:
    Sterile

Specifications