Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
red- SYLGARD 527 A & B Silicone Dielectric Gel 1 * 2 kg
Catalog # GREFSYLGARD527R2
Supplier:
GREEFF RW SILICONES LUBRICAN
Expand image
red- SYLGARD 527 A & B Silicone Dielectric Gel 1 * 2 kg
Catalog # GREFSYLGARD527R2
Supplier:
GREEFF RW SILICONES LUBRICAN