Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
test tube rack A2 for 14mm special for 70mm tubes 1 * 1 items
Catalog # BUHL0060040
Supplier:
Edmund Bühler GmbH
Expand image
test tube rack A2 for 14mm special for 70mm tubes 1 * 1 items
Catalog # BUHL0060040
Supplier:
Edmund Bühler GmbH