Order Entry
Finland
ContactUsLinkComponent
Milliflex Oasis® 250 mL funnel, 0.45 µm white gridded, mixed cellulose esters (MCE) membrane 1 * 24 items
Catalog # MILXMMHAWG224
Supplier:  Merck
Milliflex Oasis® 250 mL funnel, 0.45 µm white gridded, mixed cellulose esters (MCE) membrane 1 * 24 items
Catalog # MILXMMHAWG224
Supplier:  Merck
Related Information