Sr 3576
Bottle
Catalog # APOSBISN0280-25MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
MAPK/ERK Signalling Pathway.
Usage: Biochemical/ life science applications.
Synonyms: 3-(5-(3-m-Tolylureido)-1H-indazol-1-yl)-N-(3,4,5-trimethoxyphenyl)benzamide
- CAS Number:1164153-22-3
- Formula:C₂₇H₂₇N₅O₅
- Molecular Weight:501,53378 g/mol
Specifications
Specifications
- Pack type:Bottle



