Sr 3576
Bottle
Catalog # APOSBISN0280-10MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
MAPK/ERK Signalling Pathway.
Usage: Biochemical/ life science applications.
Synonyms: 3-(5-(3-m-Tolylureido)-1H-indazol-1-yl)-N-(3,4,5-trimethoxyphenyl)benzamide
- CAS Number:1164153-22-3
- Formula:C₂₇H₂₇N₅O₅
- Molecular Weight:501,53378 g/mol
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



