Order Entry
Finland
Orders LinkContactUsLinkComponent
1-Chloro-2-iodoethane

1-Chloro-2-iodoethane

Bottle, 5 g
Catalog # APOSOR51875-5G
Irritant
Supplier:   Apollo Scientific
1-Chloro-2-iodoethane
Catalog # APOSOR51875-5G
Supplier:  Apollo Scientific
CAS Number:  624-70-4
1-Chloro-2-iodoethane

Bottle, 5 g

CAS Number:  624-70-4
Price
158,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:624-70-4
  • Formula:C₂H₄ClI
  • MDL Number:MFCD00041453
  • Molecular Weight:190.41 g/mol
  • Boiling Pt:61…62 °C
  • Density:2.088 g/cm³ (25 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.