Order Entry
Finland
Orders LinkContactUsLinkComponent
1-Chloro-2-iodoethane

1-Chloro-2-iodoethane

Bottle, 1 g
Catalog # APOSOR51875-1G
Irritant
Supplier:   Apollo Scientific
1-Chloro-2-iodoethane
Catalog # APOSOR51875-1G
Supplier:  Apollo Scientific
CAS Number:  624-70-4
1-Chloro-2-iodoethane

Bottle, 1 g

CAS Number:  624-70-4
Price
53,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:624-70-4
  • Formula:C₂H₄ClI
  • MDL Number:MFCD00041453
  • Molecular Weight:190.41 g/mol
  • Boiling Pt:61…62 °C
  • Density:2.088 g/cm³ (25 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications