Order Entry
Finland
Orders LinkContactUsLinkComponent
Benzophenone

Benzophenone

Bottle, 500 g
Catalog # APOSOR8243-500G
Irritant
Supplier:   Apollo Scientific
Benzophenone
Catalog # APOSOR8243-500G
Supplier:  Apollo Scientific
CAS Number:  119-61-9
Benzophenone

Bottle, 500 g

CAS Number:  119-61-9
Price
72,10 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Diphenylmethanone, Diphenyl ketone
  • CAS Number:119-61-9
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00003076
  • Molecular Weight:182.22 g/mol
  • Boiling Pt:305 °C
  • Melting Pt:47…51 °C
  • Flash Pt:138 °C
  • Storage Temperature:Ambient
  • EINECS:204-337-6
  • UN:3077
  • ADR:9+EHS,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.