Order Entry
Finland
Orders LinkContactUsLinkComponent
(S)-(-)-1,2-Epoxypropane 97%

(S)-(-)-1,2-Epoxypropane 97%

Bottle, 100 g
Catalog # APOSOR480600-100G
Supplier:   Apollo Scientific
(S)-(-)-1,2-Epoxypropane 97%
Catalog # APOSOR480600-100G
Supplier:  Apollo Scientific
CAS Number:  16088-62-3
(S)-(-)-1,2-Epoxypropane 97%

Bottle, 100 g

CAS Number:  16088-62-3
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (S)-(-)-Methyloxirane, (S)-(-)-Propylene oxide, (S)-(-)-Propene oxide
  • CAS Number:16088-62-3
  • Formula:C₃H₆O
  • MDL Number:MFCD00064312
  • Molecular Weight:58.08 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.