Order Entry
Finland
Orders LinkContactUsLinkComponent
Propionic acid ≥99.7%, GPR RECTAPUR®

Propionic acid ≥99.7%, GPR RECTAPUR®

Glass bottle, 500 mL
Catalog # 87062.260
Corrosive
Flammable
Irritant
Supplier:   VWR Chemicals
Propionic acid ≥99.7%, GPR RECTAPUR®
Catalog # 87062.260
Supplier:  VWR Chemicals
CAS Number:  79-09-4
Propionic acid ≥99.7%, GPR RECTAPUR®
Propionic acid ≥99.7%, GPR RECTAPUR®

Glass bottle, 500 mL

CAS Number:  79-09-4
Price
26,20 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Boiling Pt:140…142 °C
  • Flash Pt:54 °C
  • Storage Temperature:Ambient
  • UN:3463
  • ADR:8,II
Specifications

Specification Test Results

  • Assay (GC) :
    min 99.0%
  • Identity (IR) :
    passes test
  • Density (d 20°C/4°C):
    0,993 – 0,994
  • Water (KF):
    max 0.30%
  • Appearance:
    clear liquid

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.