Order Entry
Finland
Orders LinkContactUsLinkComponent
iPlan Achromat Objective Lens
iPlan Achromat Objective Lens
iPlan achromat objective lens, 100×/0,8 oil, 1,7 m
Catalog # ZEIS4155011640000
Supplier:   ZEISS
iPlan Achromat Objective Lens
Catalog # ZEIS4155011640000
Supplier:  ZEISS
iPlan Achromat Objective Lens
iPlan achromat objective lens, 100×/0,8 oil, 1,7 m
Price
493,00 €
Log in for availability
1 * 1 Items
Product Details & Documents
ZEISS iPlan Achromat 10×/0,25 Ph1 microscope objective, infinity corrected. This lens is intended for use with the ZEISS Primostar 3 microscope.
Specifications
  • Description:
    iPlan achromat objective lens, 100×/0,8 oil
  • Seq:
    07
  • Working distance:
    1,7 m