Order Entry
Finland
ContactUsLinkComponent
Isolator glove, neoprene no staining, shape: E10 , Thickness (mm) /10 , Length (mm) 330 , Bead diameter 5mm, ambidextrous, size 8 1 * 30 items
Catalog # PIECE10330CNS8E55
Supplier:  PIERCAN
Isolator glove, neoprene no staining, shape: E10 , Thickness (mm) /10 , Length (mm) 330 , Bead diameter 5mm, ambidextrous, size 8 1 * 30 items
Catalog # PIECE10330CNS8E55
Supplier:  PIERCAN
Frequently Bought Together