Acacetin
Glass bottle, 250 mg
Catalog # ACRO465232500
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- Bioactive
Synonyms: 5,7-Dihydroxy-2-(4-methoxyphenyl)chromen-4-one, Linarisenin, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one, 4'-Methoxy-5,7-dihydroxyflavone, 5,7-Dihydroxy-4'-methoxyflavone, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4-benzopyrone, Linarigenin, Buddleoflavonol, 4'-Methoxyapigenin
- CAS Number:480-44-4
- Formula:C₁₆H₁₂O₅
- MDL Number:MFCD00016936
- Molecular Weight:284.27 g/mol
- Melting Pt:260…265 °C
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:White to light yellow to light brown Powder or crystals
- Infrared spectrum:Conforms
- Water:≤3 %
- HPLC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:250 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



